missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ DNA QuantLadders

Provide convenient, visual quantitation of DNA within ±1% accuracy

285.60 EUR - 333.00 EUR

Specifications

Buffer TE
Concentration 100 μg/mL
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Formulation Includes a vial of 6X sample loading buffer 1mL total volume 15% w/v Ficoll™ 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene Cyanol FF 30mM EDTA
View More Specs

Products 2
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
11580787
View Documents Promotion Details
Lonza™
50334
DNA QuantLadder 125 μL
333.00 EUR
Each
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
11590787
View Documents Promotion Details
Lonza™
50335
DNA Reverse QuantLadder 125 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
  • Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
  • SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
  • Store at 2° to 8°C for up to 12 months
Specifications

Specifications

TE
DNA Molecular Weight Marker/Ladder
Includes a vial of 6X sample loading buffer 1mL total volume 15% w/v Ficoll™ 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene Cyanol FF 30mM EDTA
5
2° to 8°C
100 μg/mL
Liquid
Quantitation of DNA within ±1% accuracy
125 μL
Videos
SDS
Documents

Documents

Product Certifications