missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ FlashGel™ System RNA Cassettes
Cassettes optimized for RNA. FlashGel™ System RNA Cassettes are recommended for verification and analysis of total RNA, quick checks of native RNA, and checking RNA degradation and mRNA purity.
348.00 EUR
Specifications
Concentration | 1.2% |
---|---|
Product Type | RNA Cassettes |
Separation Range | 0.5 to 9 kb |
Formulation | 1.2% Agarose |
For Use With (Application) | Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity |
Product Code | Brand | Wells | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wells | Price | Quantity & Availability | ||||||
11567855
|
Lonza™
57027 |
12 + 1-well |
348.00 EUR
Pack of 9 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||||
11577855
|
Lonza™
57028 |
16 + 1-well |
348.00 EUR
Pack of 9 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||||
Description
Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains and hazardous reagents
- Casettes fully enclose the gel, stain and running buffer
- Protect samples from contaminating RNases
- Designed for performance and purity, and are guaranteed RNase-free
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
1.2% | |
0.5 to 9 kb | |
Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity | |
Agarose | |
4.5 in. | |
11.5 cm | |
4.2 in. | |
8.4 cm | |
1.7 cm |
RNA Cassettes | |
1.2% Agarose | |
Agarose Gel, Stain and Buffer | |
9 Pack | |
2.7 in. | |
7 cm | |
10.7 cm | |
0.6 in. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- FlashGel System
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : Tetrabutylammonium hydroxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ FlashGel™ System RNA Cassettes