missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ ProSieve™ Color Protein Markers
Visible markers for use in SDS-PAGE gels
Brand: Lonza™ 50552
Additional Details : Weight : 1.00000kg
Description
- Useful for monitoring the efficiency of separation during electrophoresis
- Confirm that transfer has occurred from gel to membrane in Western blotting
ProSieve Color Protein Marker
- Proteins in the marker have been labeled with 3 different fluorescent dyes
- Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load.
- Proteins have approximate masses of 10, 15, 20, 25, 40, 50, 80, 125, and 190kDa
- Each lot is individually calibrated against a ProSieve Unstained Protein Marker
ProSieve QuadColor Protein Marker
- Proteins are individually prestained using four different dyes, producing a brightly colored ladder with an easy-to-remember pattern
- Marker is supplied ready-to-use, no heating, further dilution or addition of a reducing agent is required before use
- Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250, and 300kDa
- Apparent molecular weights are provided for each lot
Specifications
Multi | |
Lyophilized | |
Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting | |
10 to 190 kd |
DNA Molecular Weight Marker/Ladder | |
Markers have been labeled with 3 different Fluorescent dyes, Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load | |
100 μL | |
−20°C |