missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ ProSieve™ Color Protein Markers
Visible markers for use in SDS-PAGE gels
108.00 EUR - 356.00 EUR
Specifications
For Use With (Application) | Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting |
---|
Product Code | Brand | Quantity | Formulation | Range | Storage Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Range | Storage Requirements | Price | Quantity & Availability | ||
11547995
|
Lonza™
50550 |
500 μL | Markers have been labeled with 3 different Fluorescent dyes, Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load | 10 to 190 kd | −20°C |
356.00 EUR
500µL |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||
11557995
|
Lonza™
50552 |
100 μL | Markers have been labeled with 3 different Fluorescent dyes, Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load | 10 to 190 kd | −20°C |
108.00 EUR
100µL |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||
11518005
|
Lonza™
193837 |
500 μL | Prestained using four different dyes, Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250 and 300kd | 4.6 to 300 kd | −20° to 4°C |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Useful for monitoring the efficiency of separation during electrophoresis
- Confirm that transfer has occurred from gel to membrane in Western blotting
ProSieve Color Protein Marker
- Proteins in the marker have been labeled with 3 different fluorescent dyes
- Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load.
- Proteins have approximate masses of 10, 15, 20, 25, 40, 50, 80, 125, and 190kDa
- Each lot is individually calibrated against a ProSieve Unstained Protein Marker
ProSieve QuadColor Protein Marker
- Proteins are individually prestained using four different dyes, producing a brightly colored ladder with an easy-to-remember pattern
- Marker is supplied ready-to-use, no heating, further dilution or addition of a reducing agent is required before use
- Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250, and 300kDa
- Apparent molecular weights are provided for each lot
Specifications
Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting |