missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Reliant™ Precast Gels

For rapid and reproducible resolution of DNA sizes from 8bp-10kb - Lonza™ Reliant™ Precast Gels are ideal for analysis of PCR, restriction digest fragments or most cloning needs.

Brand:  Lonza™ 54907

 View more versions of this product

Product Code. 11517825

  • 428.00 EUR / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.

This item is not returnable. View return policy

Description

Description

  • Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
  • Easy to handle
  • Fit most common mini submerged electrophoresis systems
  • Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBE
1%
Precast Gel
20-well
1% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide
Agarose
Yes
0.19 in.
≤15 μL
9.5 cm
6 cm
Clear
Multiwell
300 to ≥8000 bp
Yes
DNA separation by gel electrophoresis
20 Gels
18° to 26°C
0.5 cm
3.7 in.
2.37 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.