missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Reliant™ Precast Gels
For rapid and reproducible resolution of DNA sizes from 8bp-10kb - Lonza™ Reliant™ Precast Gels are ideal for analysis of PCR, restriction digest fragments or most cloning needs.
221.00 EUR - 428.00 EUR
Specifications
Color | Clear |
---|---|
Product Type | Precast Gel |
Dnase Free | Yes |
Material | Agarose |
Quantity | 20 Gels |
Alert:
Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.
Product Code | Brand | Buffer | Wells | Formulation | Separation Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Buffer | Wells | Formulation | Separation Range | Price | Quantity & Availability | ||
11527815
|
Lonza™
54801 |
TAE | 8-well | 1% SeaKem™ Gold Agarose, TAE Buffer | 400 to ≥10,000 bp |
428.00 EUR
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11597815
|
Lonza™
54903 |
TBE | 8-well | 1% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 300 to ≥8000 bp |
428.00 EUR
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11567825
|
Lonza™
54928 |
TBE | 20-well | 4% NuSieve 3:1 Plus Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 8 to ≥1000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11507825
|
Lonza™
54905 |
TBE | 24-well | 1% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 300 to ≥8000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11557825
|
Lonza™
54927 |
TBE | 8-well | 4% NuSieve 3:1 Plus Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 8 to ≥1000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11547825
|
Lonza™
54925 |
TAE | 8-well | 4% NuSieve 3:1 Plus Agarose, TAE Buffer, 0.5μg/mL Ethidium Bromide | 20 to ≥1000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11577825
|
Lonza™
54929 |
TBE | 24-well | 4% NuSieve 3:1 Plus Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 8 to ≥1000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11527835
|
Lonza™
54938 |
TBE | 20-well | 2% SeaKem™ Gold Agarose, TBE Buffer | 100 to ≥2000 bp | N/A | |||
11537835
|
Lonza™
54939 |
TBE | 20-well | 2% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 100 to ≥2000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11547835
|
Lonza™
54944 |
TBE | 20-well | 4% NuSieve 3:1 Plus Agarose, TBE Buffer | 8 to ≥1000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11567815
|
Lonza™
54821 |
TAE | 12-well | 1% SeaKem™ Gold Agarose, TBE Buffer | 400 to ≥10,000 bp | N/A | |||
11517825
|
Lonza™
54907 |
TBE | 20-well | 1% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 300 to ≥8000 bp |
428.00 EUR
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11537815
|
Lonza™
54803 |
TAE | 8-well | 1% SeaKem™ Gold Agarose, TAE Buffer, 0.5μg/mL Ethidium Bromide | 400 to ≥10,000 bp |
428.00 EUR
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11577815
|
Lonza™
54823 |
TBE | 12-well | 4% NuSieve 3:1 Plus Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 8 to ≥1000 bp | N/A | |||
11557815
|
Lonza™
54820 |
TBE | 12-well | 1% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 300 to ≥8000 bp |
428.00 EUR
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11547815
|
Lonza™
54813 |
TBE | 24-well | 2% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 100 to ≥3000 bp |
428.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
11587815
|
Lonza™
54825 |
TBE | 12-well | 2% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | 100 to ≥3000 bp | N/A | |||
Description
- Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
- Easy to handle
- Fit most common mini submerged electrophoresis systems
- Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
Specifications
Clear | |
Yes | |
20 Gels | |
18° to 26°C | |
0.5 cm | |
3.7 in. | |
2.37 in. |
Precast Gel | |
Agarose | |
Yes | |
0.19 in. | |
≤15 μL | |
9.5 cm | |
6 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title