missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Low Iodine Diet, Fortified With Vitamins, Powder

Vitamin fortified diet, low in iodine.

810.90 EUR - 3164.10 EUR

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Powder
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11475582
View Documents
MP Biomedicals
0290220110
10 kg
810.90 EUR
10kg
Please to purchase this item. Need a web account? Register with us today!
11485582
View Documents
MP Biomedicals
0290220120
20 kg
1452.20 EUR
20kg
Please to purchase this item. Need a web account? Register with us today!
11495582
View Documents
MP Biomedicals
0290220150
50 kg
3164.10 EUR
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • This diet is low in iodine but fortified with other vitamins
  • Ideal for research on iodine deficiency diets
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Powder
Room Temperature
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.