Learn More
Cytiva Luer Connectors for Chromatography Columns
For use with the HiTrap column family of columns
54.00 EUR - 72.00 EUR
Specifications
Quantity | 2 Pack |
---|---|
Material | Polypropylene |
Column Type | Column Accesory |
Requires: Cytiva HiTrap Columns, sold separately.
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
11822180
|
Cytiva
18-1003-68 |
Tubing connecto r | Connect tubing to botto m of column |
72.00 EUR
Pack of 2 |
In Stock
Log in to see stock. |
||||
11842180
|
Cytiva
18-1027-62 |
Union | Connect a syringe to the botto m of a HiTrap column |
72.00 EUR
Pack of 2 |
In Stock
Log in to see stock. |
||||
11832180
|
Cytiva
18-1027-12 |
Union | Connect a syringe to the to p of a HiTrap column |
54.00 EUR
Pack of 2 |
In Stock
Log in to see stock. |
||||
Description
- HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
2 Pack | |
Polypropylene | |
Column Accesory |