Learn More
M3258, MedChemExpress
MedChemExpress M3258 is an orally bioavailable, potent, reversible and highly selective immunoproteasome subunit LMP7 (β5i) inhibitor.
Brand: Medchem Express HY-111790
This item is not returnable.
View return policy
Description
M3258 exerts high biochemical (IC50=3.6 nM) and cellular (IC50=3.4 nM) potency against the LMP7 subunit. M3258 shows strong antitumor efficacy in multiple myeloma xenograft models. M3258 leads to a significant and prolonged suppression of tumor LMP7 activity and ubiquitinated protein turnover and the induction of apoptosis in multiple myeloma cells.Specifications
M3258 | |
White | |
C17H20BNO5 | |
DMSO : 250 mg/mL (759.51 mM; Need ultrasonic) | |
329.16 | |
98.0% | |
Solid |
2285330-15-4 | |
Research | |
5 mg | |
O=C(N[C@@H](CC1=COC2=C1C=CC=C2)B(O)O)[C@H]3[C@@H]4CC[C@@H](O4)C3 | |
329.16 | |
Research | |
Cancer-Kinase/protease |