Learn More
M3258, MedChemExpress
MedChemExpress M3258 is an orally bioavailable, potent, reversible and highly selective immunoproteasome subunit LMP7 (β5i) inhibitor.
544.05 EUR - 823.50 EUR
Specifications
Chemical Name or Material | M3258 |
---|---|
CAS | 2285330-15-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C17H20BNO5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16484528
|
Medchem Express
HY-111790 |
10 mg |
823.50 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16417247
|
Medchem Express
HY-111790 |
5 mg |
544.05 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
M3258 exerts high biochemical (IC50=3.6 nM) and cellular (IC50=3.4 nM) potency against the LMP7 subunit. M3258 shows strong antitumor efficacy in multiple myeloma xenograft models. M3258 leads to a significant and prolonged suppression of tumor LMP7 activity and ubiquitinated protein turnover and the induction of apoptosis in multiple myeloma cells.Specifications
M3258 | |
White | |
C17H20BNO5 | |
O=C(N[C@@H](CC1=COC2=C1C=CC=C2)B(O)O)[C@H]3[C@@H]4CC[C@@H](O4)C3 | |
329.16 | |
Research | |
Cancer-Kinase/protease |
2285330-15-4 | |
Research | |
DMSO : 250 mg/mL (759.51 mM; Need ultrasonic) | |
329.16 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.