missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macro H2A.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-17197
448.80 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Macro H2A.2 Polyclonal specifically detects Macro H2A.2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Macro H2A.2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
Rabbit | |
40 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Species reactivity based on sequence identity: Bovine (100%), Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.53mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9P0M6 | |
MACROH2A2 | |
Recombinant protein encompassing a sequence within the center region of human Macro H2A.2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
55506 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macro H2A.2 Antibody, Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction