Learn More
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents
An automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples, when paired with a KingFisher purification system.
Brand:  Applied Biosystems™ A66595
 View more versions of this product
Product Code. 30054466
  • 74.50 EUR / 40mL
Explore more special offers
Description
Show More
Specifications
Specifications
Store at room temperature
Magnetic Bead
KingFisher™ Duo Prime, Flex, or Apex purification systems
Genomic DNA, Total RNA, miRNA
Sequential DNA Elution Buffer
Room Temperature
Reagent
Whole Blood, Bone Marrow
Molecular applications, including qPCR and sequencing
MagMAX™
40 mL
Flexible
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents > Sequential DNA Elution Buffer, 40 mL
For Research Use Only. Not for use in diagnostic procedures.