missing translation for 'onlineSavingsMsg'
Learn More
Magnesium carbonate basic, heavy, extra pure, Thermo Scientific Chemicals Product Code.: 15224734

Magnesium carbonate basic, heavy, extra pure, Thermo Scientific Chemicals

Product Code. 15224734
2.5 kg, Plastic Drum
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 15224734

Brand: Thermo Scientific Acros 413400025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 2.5 kg
Linear Formula MgCO3
Merck Index 15,5724
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
Formula Weight 84.31
Percent Purity 40 to 45% (MgO)
Grade Extra Pure
Packaging Plastic Drum
Chemical Name or Material Magnesium carbonate basic, heavy
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461944 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog Number413400025,413405000
Lot NumberA0452677 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number413400025,413405000
Lot NumberA0393814 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number413400025,413400100,413405000
Lot NumberA0397072 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number413400025,413400100,413405000
Lot NumberA0392359 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number413400025,413400100,413405000
Product Identifier
  • Magnesium carbonate, basic (heavy)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium carbonate basic, heavy, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.