missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
1 kg
Description
- Molecular Formula: MgCl2 • 6H2O
- Molar Mass: 203.30g/mol
- CAS: 7791-18-6
- EIN ECS No.: 232-094-6
Specifications
Specifications
Melting Point | ∼117°C (decomposition) |
CAS | 7791-18-6 |
pH | 5.0–6.5 (5%, water) |
Quantity | 1 kg |
Molecular Formula | Cl2H12MgO6 |
MDL Number | MFCD00149781 |
InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
IUPAC Name | magnesium(2+) hexahydrate dichloride |
Molecular Weight (g/mol) | 203.30 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Magnesium Chloride Hexahydrate, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction