Learn More
Invitrogen™ MagniSort™ Mouse CD4 Positive Selection Kit
Designed for the magnetic separation of CD4+ cells by positive selection. It has been optimized for the isolation of CD4+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD4 antibody and streptavidin-coated magnetic beads.
Brand:  Invitrogen™ 8802-6841-74
Product Code. 15521257
  • 804.00 EUR / 200 tests
Explore more special offers
Alert:
Contains Sodium azide
Includes:
  • MagniSort Anti-Mouse CD4 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A
Description
Description
  • CD4+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD4+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD4, clones RM4-5 or GK1.5
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4+
200 tests
Mouse
Positive Selection Kit
200 tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528509 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8802-6841-74
Lot Number2618909 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8802-6841-74
Lot Number2829454 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number8802-6841-74
Lot Number2913935 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number8802-6841-74
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ MagniSort™ Mouse CD4 Positive Selection Kit > MagniSort Mouse CD4 Positive Selection Kit
For Research Use Only. Not for use in diagnostic procedures.