Learn More
Value Plastics Male Luer Integral Lock Ring to Plug Adapters
Seal luer ports with a secure and leak-proof connection
Brand:  Value Plastics 50120-09
 View more versions of this product
Product Code. 30054009
  • 17.34 EUR / Pack of 25
Description
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism
Specifications
Specifications
Non-DEHP;Non-Phthalate;RoHS;FDA
Male Luer Integral Lock Ring, Plug, Closed at Grip
Polycarbonate