Learn More
Value Plastics Male Luer Integral Lock Ring to Plug Adapters
Seal luer ports with a secure and leak-proof connection
13.83 EUR - 21.98 EUR
Products 3
Product Code Brand Material (Seal) Price Quantity & Availability  
Product Code Brand Material (Seal) Price Quantity & Availability  
30054073
View Documents
Value Plastics
50120-74
Nylon
13.83 EUR
Pack of 25
30054009
View Documents
Value Plastics
50120-09
Polycarbonate
17.34 EUR
Pack of 25
30053291
View Documents
Value Plastics
50120-75
polypropylene
21.98 EUR
Pack of 25
Description
Description
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism