Learn More
Value Plastics Male Luer Integral Lock Ring to Plug Adapters
Seal luer ports with a secure and leak-proof connection
Brand: Value Plastics 50120-09
This item is not returnable.
View return policy
Description
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanismSpecifications
Non-DEHP;Non-Phthalate;RoHS;FDA | |
Male Luer Integral Lock Ring, Plug, Closed at Grip | |
Polycarbonate |