missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAPK7 (ERK5), GST Tag Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ A30502
This item is not returnable.
View return policy
Description
MAPK7 (ERK5) serine/threonine kinase is a member of the MAP kinase family and is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. MAPK7 is activated by mitogen-activated protein kinase 5 (MAP2K5/MEK5) and is involved in downstream signaling processes of various receptors. In response to extracelluar signals, MAPK7 translocates to the cell nucleus where it regulates gene expression by phosphorylating and activating different transcription factors. Gene deletion of MAPK7 in mice results in defective blood vessel and cardiac development leading to embryonic lethality. MAPK7 has been shown to be critical for endothelial function and maintenance of blood vessel integrity. Four alternatively spliced transcript variants of this gene encoding two distinct isoforms have been reported. ERK5 is expressed in many adult tissues, abundantly in heart, placenta, lung, kidney and skeletal muscle, but is not detectable in liver.
Specifications
Q13164 | |
Kinase Assay | |
5598 | |
Human MAPK7 (ERK5), GST Tag | |
Big MAP kinase 1; BMK1; BMK-1; BMK1 kinase; BNhp-1; BNhp1 kinase; ERK4; ERK5; ERK-5; Erk5-T; extracellular signal regulated kinase 5; extracellular signal-regulated kinase 5; extracellular-signal-regulated kinase 5; MAP kinase 7; MAPK 7; Mapk7; mitogen activated protein kinase 7; mitogen-activated protein kinase 7; PRKM7 | |
MAP Kinases | |
Human | |
GST-tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Insect cells | |
Full Length, Recombinant, Active |
See Label | |
50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
114.7 kDa | |
10 μg | |
ERK5 | |
Unconjugated | |
Recombinant | |
Full length | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human MAPK7 (ERK5), GST Tag Recombinant Protein > 10 μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction