missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPT Monoclonal Antibody (OTI13E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809192S

This item is not returnable.
View return policy
Description
MAPT Monoclonal antibody specifically detects MAPT in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
MAPT | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NP77 | |
Mouse | |
Affinity chromatography | |
RUO | |
29101 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI13E11 | |
Unconjugated | |
Ssu72 | |
AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
Full length human recombinant protein of human SSU72 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MAPT Monoclonal Antibody (OTI13E11), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction