missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MATRIX PCR Attachment

The ideal attachment design ensures an optimal temperature transfer and thus guarantees valid results.

Brand:  IKA™ 0030000857

 View more versions of this product

Product Code. 16268691

  • 708.00 EUR / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The thermoshakers MATRIX Orbital Delta plus and MATRIX Delta plus automatically recognize, which attachment is connected to the device via an interface in the locking mechanism. The attachment can be changed quickly and easily with an ergonomic lever. Suitable for: MATRIX Orbital Delta plus MATRIX Delta plus
Specifications

Specifications

Reaction Vessels
150 x 141 x 42 mm
MATRIX Orbital Delta Plus, MATRIX Delta Plus
PCR 384 Attachment for 384-Well-PCR Plates
Samples with even the smallest volume, such as blood samples, pharmaceutical agents, ELISA assays/DNA/RNA samples
384
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.