missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ MATRIX PCR Attachment
The ideal attachment design ensures an optimal temperature transfer and thus guarantees valid results.
661.00 EUR - 708.00 EUR
Specifications
Holds | Reaction Vessels |
---|---|
Dimensions (L x W x H) | 150 x 141 x 42 mm |
For Use With (Application) | Samples with even the smallest volume, such as blood samples, pharmaceutical agents, ELISA assays/DNA/RNA samples |
For Use With (Equipment) | MATRIX Orbital Delta Plus, MATRIX Delta Plus |
Product Code | Brand | Product Type | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
16258691
|
IKA™
0030000853 |
Attachment for PCR Reaction Vessels/96-Well-PCR Plate | 150 x 141 x 42 mm |
661.00 EUR
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
16268691
|
IKA™
0030000857 |
PCR 384 Attachment for 384-Well-PCR Plates | 150 x 141 x 42 mm |
708.00 EUR
Each |
In Stock
Log in to see stock. |
||||
Description
The thermoshakers MATRIX Orbital Delta plus and MATRIX Delta plus automatically recognize, which attachment is connected to the device via an interface in the locking mechanism. The attachment can be changed quickly and easily with an ergonomic lever. Suitable for: MATRIX Orbital Delta plus MATRIX Delta plusSpecifications
Reaction Vessels | |
150 x 141 x 42 mm | |
Samples with even the smallest volume, such as blood samples, pharmaceutical agents, ELISA assays/DNA/RNA samples | |
MATRIX Orbital Delta Plus, MATRIX Delta Plus |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title