Learn More
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Click to view available options
Color:
Black
Blue
Colorless
Gray
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Screw Cap
Screw Cap Insert
Screw Cap Tray
Screw Cap Tray, Empty
Unit Size:
Pack of 480
Pack of 5
Pack of 500
Descripción
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Superior Cap Design
- Gasketed caps ensure critical sample protection over the storage lifetime
- Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
- Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
- ScrewTop caps are compatible with storage down to -196ºC
Cap Options Simplify Sample Identification
- Cap trays feature standard microplate format compatible with automated equipment
- Caps are available in seven color options to enable sample identification
- Color inserts snap into cap for a secure fit; inserts are removable
- Inserts are available in six colors
All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins
Especificaciones
Especificaciones
| Product Type | Screw Cap |
| Material | Polypropylene |
| Color | Orange |
| Packaging | Bulk, 500 Caps |
| Sterility | Sterile |
| For Use With (Equipment) | ScrewTop Tube |
| Fit for Purpose | Sterile and PCR Ready |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido