missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin (Mesothelial Marker) Mouse, Clone: MSLN/2131, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 10232-MSM1-P0
This item is not returnable.
View return policy
Specifications
Mesothelin (Mesothelial Marker) | |
Cell Surface and Secreted | |
MSLN/2131 | |
Unconjugated | |
Mouse | |
40kDa (Mature form); 69kDa (Precursor) | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
10232 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment (apprx. aa 273-407) of human Mesothelin (MSLN) protein | |
CAPAN-2 or HeLa cells. Lung Mesothelioma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |