missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin (Mesothelial Marker) Mouse, Clone: MSLN/2131, enQuireBio™
Mouse Monoclonal Antibody
199.00 EUR - 458.00 EUR
Specifications
Antigen | Mesothelin (Mesothelial Marker) |
---|---|
Clone | MSLN/2131 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949707
|
EnquireBio
10232-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959707
|
EnquireBio
10232-MSM1-P1 |
100 μg |
458.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Spezifikation
Mesothelin (Mesothelial Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment (apprx. aa 273-407) of human Mesothelin (MSLN) protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
40kDa (Mature form); 69kDa (Precursor) |
MSLN/2131 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
10232 | |
IgG2b κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface and Secreted | |
CAPAN-2 or HeLa cells. Lung Mesothelioma. |