Learn More
Jenway™ Meter Accessory and Replacement Part, Micro Volume Conductivity Probe
Designed to optimize the performance of your Jenway meter.
Brand:  Jenway™ 99959-72
Product Code. 15587019
  • 690.00 EUR / Each
Estimated Shipment: 10-06-2025
Description
  • For use with Jenway Meter
  • Easy to install
  • Accessory
Specifications
Specifications
1
Jenway Meter
Glass
7-pin
Micro Volume Conductivity Probe
0 to 150 mS/cm
Product Title
Jenway™ Meter Accessory and Replacement Part, Micro Volume Conductivity Probe > Micro Volume Conductivity Probe