Learn More
Methacycline hydrochloride, MedChemExpress
MedChemExpress Methacycline hydrochloride is a tetracycline antibiotic and can inhibits bacterial protein synthesis. Methacycline hydrochloride is a potent epithelial-mesenchymal transition (EMT) inhibitor. Methacycline hydrochloride blocks EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. Methacycline hydrochloride is an antimicrobial and has the potential for pulmonary fibrosis.
Brand:  Medchem Express HY-B0449
Encompass_Preferred
 View more versions of this product
Product Code. 16411426
  • 137.70 EUR / 200mg
Explore more special offers
Description
Description
Methacycline hydrochloride is a tetracycline antibiotic and can inhibits bacterial protein synthesis. Methacycline hydrochloride is a potent epithelial-mesenchymal transition (EMT) inhibitor. Methacycline hydrochloride blocks EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. Methacycline hydrochloride is an antimicrobial and has the potential for pulmonary fibrosis.
Specifications
Specifications
Methacycline hydrochloride
Yellow
C22H23ClN2O8
DMSO : 50 mg/mL (104.41 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (13.93 mM; Need ultrasonic)
478.88
99.53%
Solid
3963-95-9
Research
200 mg
O[C@@](C(O)=C(C(C1=C2C=CC=C1O)=O)[C@](C2=C)([H])[C@@H]3O)(C(C(C(N)=O)=C4O)=O)[C@]3([H])[C@@H]4N(C)C.Cl
478.88
Research
COVID-19-immunoregulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Methacycline hydrochloride, MedChemExpress