Learn More
Methimazole-d3, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
CAS | 1160932-07-9 |
Packaging | Glass Bottle |
Quantity | 10 mg |
For Use With (Application) | Steroids and Hormones |
Physical Form | Solid |
Grade | Analytical Standard |
Product Content Correction
Product Title
Methimazole-d3, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction