missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1H-indazole-6-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl 1H-indazole-6-carboxylate is used in organic synthesis.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Methyl 1H-indazole-6-carboxylate is used in organic synthesis.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 142°C to 144°C |
Quantity | 250 mg |
Solubility Information | Soluble in water. |
Formula Weight | 176.17 |
Percent Purity | 95% |
Chemical Name or Material | Methyl 1H-indazole-6-carboxylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl 1H-indazole-6-carboxylate, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction