missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-chloro-3-iodobenzoate, 98+%, Thermo Scientific Chemicals

81.90 EUR - 299.00 EUR

Chemical Identifiers

CAS 289039-84-5
Molecular Formula C8H7ClINO2
Molecular Weight (g/mol) 311.503
MDL Number MFCD00673017
InChI Key VTDNWMZYPVQPCN-UHFFFAOYSA-N
PubChem CID 7020924
IUPAC Name methyl 2-amino-5-chloro-3-iodobenzoate
SMILES COC(=O)C1=CC(=CC(=C1N)I)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359016
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L20188.06
5 g N/A N/A N/A
11369016
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L20188.14
25 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-amino-5-chloro-3-iodobenzoate, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.