missing translation for 'onlineSavingsMsg'
Learn More
Methyl 3-amino-6-bromopyrazine-2-carboxylate, 97%, Thermo Scientific™ Product Code.: 15410279

Methyl 3-amino-6-bromopyrazine-2-carboxylate, 97%, Thermo Scientific™

Product Code. 15410279
5 g
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15410279

Brand: Thermo Scientific Alfa Aesar H63537.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6966-01-4
Molecular Formula C6H6BrN3O2
Molecular Weight (g/mol) 232.04
MDL Number MFCD00834964
InChI Key CNXSIRHOIFRMOB-UHFFFAOYSA-N
Synonym methyl3-amino-6-bromopyrazine-2-carboxylate, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 2-amino-5-bromo-3-methoxycarbonyl pyrazine, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, pubchem22386
PubChem CID 227251
IUPAC Name methyl 3-amino-6-bromopyrazine-2-carboxylate
SMILES COC(=O)C1=NC(=CN=C1N)Br

Specifications

Melting Point 172°C to 177°C
Color White to Yellow
Quantity 5 g
Formula Weight 232.03
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-amino-6-bromopyrazine-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3-amino-6-bromopyrazine-2-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.