missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-nitrobenzoate, 97%, Thermo Scientific Chemicals

41.51 EUR - 229.14 EUR

Chemical Identifiers

CAS 3987-92-6
Molecular Formula C8H8N2O4
Molecular Weight (g/mol) 196.162
MDL Number MFCD00017562
InChI Key HNTLUEZVPLRQEV-UHFFFAOYSA-N
Synonym methyl 4-amino-3-nitrobenzenecarboxylate, benzoic acid, 4-amino-3-nitro-, methyl ester, 4-amino-3-nitro-benzoic acid methyl ester, 4-methoxycarbonyl-2-nitroaniline, 4-amino-3-nitromethylbenzoate, methyl 3-nitro-4-aminobenzoate, 2-amino-5-methoxycarbonyl nitrobenzene, 3-nitro-4-aminobenzoic acid methyl ester, 4-amino-3-nitrobenzoicacidmethylester, acmc-209j8j
PubChem CID 3941008
IUPAC Name methyl 4-amino-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15437287
SDS View Documents
Thermo Scientific Alfa Aesar
H34348.03
1 g
41.51 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
15447287
SDS View Documents
Thermo Scientific Alfa Aesar
H34348.09
10 g
229.14 EUR
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4-amino-3-nitrobenzoate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.