missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Density | 1.524 |
Boiling Point | 83°C to 85°C (13 mmHg) |
Flash Point | 91°C (195°F) |
Linear Formula | BrCH2CH=CHCO2CH3 |
Refractive Index | 1.501 |
Quantity | 10 g |
UN Number | UN1760 |
Beilstein | 1745755 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 179.01 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction