Learn More
Methyl cinnamate, predominantly trans, 99%, Thermo Scientific Chemicals
Description
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 34°C to 37°C |
Density | 1.042 |
Boiling Point | 262°C to 264°C |
Flash Point | 123°C (253°F) |
Assay Percent Range | 99% |
Refractive Index | 1.577 |
Quantity | 500 g |
Beilstein | 386468 |
Merck Index | 14,2299 |
Solubility Information | Insoluble in water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10248262 | Certificate of Analysis | 16/10/2023 | A15975.22,A15975.36,A15975.0E |
G8182C | Certificate of Analysis | 23/11/2021 | A15975.22,A15975.36,A15975.0E |
10230484 | Certificate of Analysis | 23/11/2021 | A15975.22,A15975.36,A15975.0E |
10003607 | Certificate of Analysis | 23/11/2021 | A15975.22,A15975.36,A15975.0E |
10220430 | Certificate of Analysis | 23/11/2021 | A15975.22,A15975.36,A15975.0E |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.