Learn More
Mettler Toledo™ DL31 Karl Fischer™ Titrator Replacement Parts: Electrodes
Replacement parts for DL31 Karl Fischer titrators.
Brand:  Mettler Toledo™ 89598
 View more versions of this product
Product Code. 12734436
  • 764.00 EUR / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Description
Combined platinum ring redox electrode with a ceramic frit for redox titrations with change of the pH value.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Combined Platinum Ring Electrode
Ceramic frit
149.5 mm
0°C to 80°C
For redox titrations
5.8 in.
0°F to 176°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ DL31 Karl Fischer™ Titrator Replacement Parts: Electrodes > Redox electrode