Learn More
Mettler Toledo™ Paper Rolls for P50 Line Thermal Printers
For use with P50 Line Thermal Printer.
Brand: Mettler Toledo™ 30094723
73.19 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
Thermal Paper Roll
- Accessory for P-56RUE and P-58RUE Thermal printers
- White in color
- Up to 25 years durability
- Ten pieces
Specifications
P-56RUE and P-58RUE Thermal Printers | |
10 Pieces | |
Thermal Paper Roll |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now