Learn More
Biochrom Micro-Volume Cuvette Bubble Viewer
Micro-Volume Cuvette Bubble Viewer helps you determine if air pockets are present in your sample before taking a measurement.
Brand: Biochrom 80-3006-26
This item is not returnable.
View return policy
Description
- Helps to make sure your sample is ready for measurement
Specifications
Micro-Volume Cuvette Bubble Viewer for 15 mm Beam Height Cuvettes | |
Reusable | |
Microvolume cuvettes |
Microcell Holder | |
15 mm Beam Height Cuvettes | |
Square |
Product Content Correction
Product Title
For research use only.
Spot an opportunity for improvement?Share a Content Correction