Learn More
Biochrom Micro-Volume Cuvette Bubble Viewer
Micro-Volume Cuvette Bubble Viewer helps you determine if air pockets are present in your sample before taking a measurement.
229.00 EUR
Description
- Helps to make sure your sample is ready for measurement
Specifications
Microcell Holder | |
Reusable |
For research use only.