Learn More
Biochrom Micro-Volume Cuvette Bubble Viewer
Micro-Volume Cuvette Bubble Viewer helps you determine if air pockets are present in your sample before taking a measurement.
229.00 EUR
Specifications
Type Microcell Holder
Disposable Reusable
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15801232
View Documents
Biochrom
80-3006-27
8.5 mm Beam Height Cuvettes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15891222
View Documents
Biochrom
80-3006-26
15 mm Beam Height Cuvettes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Helps to make sure your sample is ready for measurement
Specifications
Specifications
Microcell Holder
Reusable
Videos
SDS
Documents
Documents
Product Certifications
For research use only.