Learn More
Heathrow Scientific Microcentrifuge Tubes, Natural
Microcentrifuge tubes have attached flat caps for easy one-handed use.
34.45 EUR - 53.20 EUR
Specifications
Cap Style | Flat |
---|---|
Closure Color | Natural |
Closure Material | Polypropylene |
Description | Microcentrifuge Tube |
Color | Natural |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
16300776
|
Heathrow Scientific
HS4325 |
1.5 mL |
34.45 EUR
Pack of 1000 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
16310776
![]() |
Heathrow Scientific
HS4422 |
0.5 mL |
35.70 EUR
Pack of 1000 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
16320776
![]() |
Heathrow Scientific
HS4423 |
0.2 mL |
53.20 EUR
Pack of 1000 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
Description
- 100% polypropylene
- Frosted writing area and a syringe pierceable cap
- Autoclavable
- Disposable
Specifications
Flat | |
Polypropylene | |
Natural | |
Non-sterile |
Natural | |
Microcentrifuge Tube | |
Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Heathrow Scientific Microcentrifuge Tubes, Natural