Learn More
Fisherbrand™ Microcentrifuges, Micro 17/17R
GREENER_CHOICE
Adjustable 99-minute timer and quick-spin feature for short runs.
Brand:  Fisherbrand™ 75002462
3470.55 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 11526873
  • 5130.00 EUR / Each
Description
  • Capacity: 48mL (24 x 1.5/2mL tubes)
  • Maximum speed/RCF: 13300rpm/17000xg
  • Adjustable 99 minute timer and quick-spin feature for short runs
  • Supplied with click seal rotor (24 place 1.5/2mL) and biocontainment lid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
17,000 x g
Yes
13,300 rpm
Microcentrifuge Micro 17R, refrigerated
Videos
  • 16574_Fisherbrand - POTM - February EN
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Microcentrifuges, Micro 17/17R (EN)
    SDS
    Documents Certificates
    View accessory products
    Special Offers
    Special Offers
    • Thermo Scientific Sorvall , Fisherbrand , Thermo Scientific
      Unlock special pricing on top centrifuges. Make us an offer today and upgrade your lab equipment. Limited time only!
      Promo Code: 21629
      Expires2025-12-31
    • Multiple Manufacturers
      Browse exclusive prices for our academic customers
      Promo Code: 25081
      Expires2025-12-31
    • Multiple Manufacturers
      Use promotion code 24138 to get amazing savings on products for your analysis
      Promo Code: 24138
      Expires2025-04-30
    • Multiple Manufacturers
      Save up to 94% on Teaching Lab Essentials
      Promo Code25721
      Expires2025-05-01
      New Offer
      Save Now
    Product Content Correction
    Product Title
    Fisherbrand™ Microcentrifuges, Micro 17/17R > Microcentrifuge Micro 17R, refrigerated