missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Micropipette Filter

Vitlab™ Micropipette Filter for use with5mL and 10mL micropipettes.

Brand:  Vitlab™ 1672012

 View more versions of this product

Product Code. 15791986

  • 12.80 EUR / Pack of 25
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Filter for pipette
  • Accessory
Specifications

Specifications

10 mL
Pipette Accessory
Micropipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers