missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Micropipette Filter

Vitlab™ Micropipette Filter for use with5mL and 10mL micropipettes.

7.50 EUR - 12.80 EUR

Specifications

Type Pipette Accessory
For Use With (Equipment) Micropipette
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15781986
View Documents Promotion Details
Vitlab™
1672010
7.50 EUR
Pack of 25
View alternative products
Save Up to 
»
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
 
15791986
View Documents Promotion Details
Vitlab™
1672012
10 mL
12.80 EUR
Pack of 25
View alternative products
Save Up to 
»
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
 
Description

Description

  • Filter for pipette
  • Accessory
Specifications

Specifications

Pipette Accessory
Micropipette
Videos
SDS
Documents

Documents

Product Certifications