Learn More
Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Integrate these microplate carriers with the microplate swinging bucket rotor.
Brand: Thermo Scientific™ 75003625
1086.35 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Integrate Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor to add flexibility to the swinging-bucket rotor.
Includes:
Sealed carriers include trays and lids (set of 2). Replacement O-rings for lids are offered as a set of four, with grease.
Specifications
Microplate Carrier | |
Sealed Microplate Carriers | |
Biocontainment certified by CAMR™, HPA, Porton Down, U.K. |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%