Learn More
Ohaus™ Microtube Foam Insert
OHAUS™ Microtube Foam Insert is suitable for use with vortex mixers.
Brand: Ohaus™ 30400217
39.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
38 Mircotubes | |
Foam | |
38 |
Foam Insert | |
Foam Insert (38) 1.5/2.0 mL Microtubes |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Ohaus™ Microtube Foam Insert > Material: Polyethylene Foam
Spot an opportunity for improvement?Share a Content Correction