Learn More
Brady™ MiniMark™ Polyester Name and Rating Plates Labels
Identify your Asset and Rating Plates with these metallic looking MiniMark™ Polyester Name and Rating Plates Labels.
Brand:  Brady™ 710090
 View more versions of this product
Product Code. 11771795
  • 332.00 EUR / 1 box
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Description
  • Identify your Asset and Rating Plates with these metallic looking labels.
Specifications
Specifications
ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognised
MiniMark Name and Rating Plates Labels
White
Polyester
Labels
MiniMark Printer
-80°C
1240 Pk.
-112°F to +230°F
0.0028 in.
28.22 mm
1 in.
2 in.
MNK-BPT-17-8423
Permanent Acrylic, Permanent
Satin
Thermal Transfer
Asset tracking, General Identification, Laboratory labelling, Circuit board labelling, Warehouse marking, Barcode labelling
1
1240 Labels/Box
50.8 x 25.4 mm (W x H)
-80°C to +110°C
0.071 mm
25.4 x 50.8 mm
25.4 mm
50.8 mm