Learn More
Brady™ MiniMark™ Polyester Name and Rating Plates Labels
Identify your Asset and Rating Plates with these metallic looking MiniMark™ Polyester Name and Rating Plates Labels.
184.00 EUR - 358.00 EUR
Specifications
Certifications/Compliance | ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognised |
---|---|
Description | MiniMark Name and Rating Plates Labels |
Adhesive Type | Permanent Acrylic, Permanent |
Color | White |
Finish | Satin |
Product Code | Brand | Width (Metric) | HeightMetric | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | HeightMetric | Price | Quantity & Availability | ||||
11731825
|
Brady™
710086 |
25.4 mm | N/A | ||||||
11761825
|
Brady™
710087 |
38.1 mm |
332.00 EUR
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
11771795
|
Brady™
710090 |
50.8 mm |
332.00 EUR
1 box |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
11791785
|
Brady™
710088 |
69.85 mm |
358.00 EUR
1 box |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
Description
- Identify your Asset and Rating Plates with these metallic looking labels.
Specifications
ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognised | |
Permanent Acrylic, Permanent | |
Satin | |
Thermal Transfer | |
Asset tracking, General Identification, Laboratory labelling, Circuit board labelling, Warehouse marking, Barcode labelling | |
-80°C | |
-80°C to +110°C | |
0.071 mm |
MiniMark Name and Rating Plates Labels | |
White | |
Polyester | |
Labels | |
MiniMark Printer | |
-112°F to +230°F | |
0.0028 in. |