Learn More
Brady™ MiniMark™ Pre-Printed Paper Labels
These MiniMark™ Pre-Printed Paper Labels are supplied with pre-printed red diamonds and are ready for you to print the required warning.
Brand: Brady™ 710586
This item is not returnable.
View return policy
Description
- Supplied with pre-printed red diamonds.
- Ready for you to print the required warning.
- Compliant to the latest regulations.
Specifications
MNK-CLP-105x155-L4B-B7610-0.2 | |
Permanent Acrylic, Permanent | |
Matte | |
Thermal Transfer | |
Hazard Communication Identification | |
-40°C | |
200 Pk. | |
MiniMark | |
-40°C to +50°C | |
0.081 mm | |
4.13 in. | |
6.10 in. |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | |
Red on White | |
Paper | |
Labels | |
MiniMark Printer | |
200 Labels/Box | |
Diamond | |
-40°F to +122°F | |
0.0032 in. | |
105 x 155 mm | |
105 mm | |
155 mm |
Product Content Correction
Product Title
Brady™ MiniMark™ Pre-Printed Paper Labels > MiniMark™ Pre-Printed Paper Labels; Dimensions (W x H): 155 x 105 mm; 4 Diamonds, Diamond Shape
Spot an opportunity for improvement?Share a Content Correction