Learn More
Invitrogen™ MitoTracker™ Red FM Dye, for flow cytometry
Mitotracker™ dyes for flow cytometry are cell permeant reagents that label mitochondria in live cells. Provided at an optimized concentration with protocols for flow cytometry to allow for more precise analysis with both conventional and spectral flow cytometry.
Brand:  Invitrogen™ M46751
133.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Product Code. 17511655
  • 200.00 EUR / 100 tests
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
  • Cell permeant reagents that label mitochondria in live cells
  • Dyes are provided at an optimized concentration with protocols for flow cytometry to allows for more precise analysis with both conventional and spectral flow cytometry
  • Simply incubate cells in the recommended dye concentration for flow cytometry, which passively diffuses across the plasma membrane and accumulates in active mitochondria
  • Bright colors allow for easy detection of mitochondria using flow cytometry without the need for special filters
  • Dyes are consistent, reliable, and stable allowing for the generation of high-quality data
  • Optimized for flow cytometry use with all essential reagents for labeling mitochondria in a single kit
  • Dissolve a vial of dye in anhydrous DMSO to provide enough dye for 20 assays
  • A range of available colors are detected with standard flow cytometry filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry
Dye
5 vials x 20 tests
Room Temperature
5 x 20 tests
100 tests
RUO
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3026199 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog NumberM46751
Lot Number2920609 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog NumberM46751
Lot Number2747517 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog NumberM46751
Lot Number2585751 Certificate TypeCertificate of Analysis Date10/07/2023 Catalog NumberM46751
Lot Number2541543 Certificate TypeCertificate of Analysis Date11/10/2022 Catalog NumberM46751
Product Title
Invitrogen™ MitoTracker™ Red FM Dye, for flow cytometry > 100 tests