missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ ML 418

Kir 7.1 inhibitor. Also inhibits Kir 6.2

219.00 EUR - 787.00 EUR

Specifications

Chemical Name or Material Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate
CAS 1928763-08-9
Molecular Formula C19H24CIN3O3
Purity 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18715946
View Documents
Tocris™
6889/10
10 mg
219.00 EUR
10mg
Please to purchase this item. Need a web account? Register with us today!
18709776
View Documents
Tocris™
6889/50
50 mg
787.00 EUR
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Kir7.1 inhibitor (IC50 = 0.31 μM)
  • Also inhibits Kir6.2 with similar potency
  • Displays >17-fold selectivity for Kir7.1 over Kir1.1, Kir2.1, Kir2.2, Kir2.3, Kir3.1/3.2 and Kir4.1
  • Active in vivo.
Specifications

Specifications

Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate
1928763-08-9
C19H24CIN3O3
98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris™ ML 418

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.