Learn More
MM-102 TFA, MedChemExpress
MedChemExpress MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor, achieves IC50 = 2.4 nM with an estimated Ki 200 times more potent than the ARA peptide.

83.70 EUR - 1271.70 EUR
Specifications
Chemical Name or Material | MM-102 TFA |
---|---|
CAS | 1883545-52-5 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C37H50F5N7O6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16481267
![]() |
Medchem Express
HY-12220A |
50 mg |
1271.70 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16467757
![]() |
Medchem Express
HY-12220A |
5 mg |
183.60 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430156
![]() |
Medchem Express
HY-12220A |
2 mg |
83.70 EUR
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16405038
![]() |
Medchem Express
HY-12220A |
10 mg |
301.05 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor, achieves IC50 = 2.4 nM with an estimated Ki 200 times more potent than the ARA peptide.Specifications
MM-102 TFA | |
White | |
C37H50F5N7O6 | |
DMSO : ≥ 100 mg/mL (127.58 mM) | |
783.83 | |
98.89% | |
Solid |
1883545-52-5 | |
Research | |
HMTase Inhibitor IX TFA | |
O=C(C1(NC([C@@H](NC(C(NC(C(C)C)=O)(CC)CC)=O)CCCNC(N)=N)=O)CCCC1)NC(C2=CC=C(F)C=C2)C3=CC=C(F)C=C3.FC(F)(F)C(O)=O | |
783.83 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.